Assay Details
| Gene Symbol: LOC4331462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776517.2 | XP_015632003.1 | 6 - 7 | 533 | 72 | |
| GenBank mRNA | AK065004.1 | - | 6 - 7 | 527 | 72 | |
| AK121790.1 | - | 6 - 7 | 527 | 72 | ||
More Information
Set Membership: |
|

LOC4331462
Genomic Map