Assay Details
Gene Symbol: LOC4331669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776984.2 | XP_015632470.1 | 2 - 3 | 1130 | 102 | |
GenBank mRNA | AK103466.1 | - | 2 - 3 | 1104 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
