Assay Details
| Gene Symbol: LOC4332184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772426.2 | XP_015627912.1 | 3 - 4 | 904 | 88 | |
| GenBank mRNA | AY187941.1 | - | 3 - 4 | 728 | 88 | |
More Information
Set Membership: |
|

LOC4332184
Genomic Map