Assay Details
| Gene Symbol: LOC4332457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774877.2 | XP_015630363.1 | 1 - 2 | 374 | 76 | |
| XM_015774878.2 | XP_015630364.1 | 1 - 2 | 372 | 76 | ||
| GenBank mRNA | AK070169.1 | - | 1 - 2 | 355 | 76 | |
More Information
Set Membership: |
|

LOC4332457
Genomic Map