Assay Details
Gene Symbol: LOC4332457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774877.2 | XP_015630363.1 | 3 - 4 | 567 | 152 | |
XM_015774878.2 | XP_015630364.1 | 3 - 4 | 565 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
