Assay Details
| Gene Symbol: LOC4332570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773039.2 | XP_015628525.1 | 2 - 3 | 406 | 57 | |
| GenBank mRNA | AK059801.1 | - | 2 - 3 | 333 | 57 | |
| AK066250.1 | - | 2 - 3 | 412 | 57 | ||
| AK069897.1 | - | 2 - 3 | 375 | 57 | ||
| AK104186.1 | - | 2 - 3 | 377 | 57 | ||
More Information
Set Membership: |
|

LOC4332570
Genomic Map