Assay Details
| Gene Symbol: LOC4332570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773039.2 | XP_015628525.1 | 3 - 4 | 478 | 69 | |
| GenBank mRNA | AK059801.1 | - | 3 - 4 | 405 | 69 | |
| AK066250.1 | - | 3 - 4 | 484 | 69 | ||
| AK069897.1 | - | 3 - 4 | 447 | 69 | ||
| AK104186.1 | - | 3 - 4 | 449 | 69 | ||
More Information
Set Membership: |
|

LOC4332570
Genomic Map