Assay Details
| Gene Symbol: LOC4332976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774692.2 | XP_015630178.1 | 1 - 2 | 268 | 103 | |
| GenBank mRNA | AK069332.1 | - | 1 - 2 | 264 | 103 | |
More Information
Set Membership: |
|

LOC4332976
Genomic Map