Assay Details
| Gene Symbol: LOC4333171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775365.1 | XP_015630851.1 | 10 - 11 | 3503 | 133 | |
| XM_015775366.2 | XP_015630852.1 | 10 - 11 | 3481 | 133 | ||
| GenBank mRNA | AK065099.1 | - | 10 - 11 | 3332 | 133 | |
| AK067814.1 | - | 10 - 11 | 3453 | 133 | ||
| AK103000.1 | - | 7 - 8 | 2938 | 133 | ||
More Information
Set Membership: |
|

LOC4333171
Genomic Map