Assay Details
| Gene Symbol: LOC4333171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775365.1 | XP_015630851.1 | 11 - 12 | 3639 | 111 | |
| XM_015775366.2 | XP_015630852.1 | 11 - 12 | 3617 | 111 | ||
| GenBank mRNA | AK065099.1 | - | 11 - 12 | 3468 | 111 | |
| AK067814.1 | - | 11 - 12 | 3589 | 111 | ||
| AK073276.1 | - | 4 - 5 | 818 | 111 | ||
| AK103000.1 | - | 8 - 9 | 3074 | 111 | ||
More Information
Set Membership: |
|

LOC4333171
Genomic Map