Assay Details
| Gene Symbol: LOC4333171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775365.1 | XP_015630851.1 | 12 - 13 | 3867 | 75 | |
| XM_015775366.2 | XP_015630852.1 | 12 - 13 | 3845 | 75 | ||
| GenBank mRNA | AK065099.1 | - | 12 - 13 | 3696 | 75 | |
| AK067814.1 | - | 12 - 13 | 3817 | 75 | ||
| AK073276.1 | - | 5 - 6 | 75 | |||
| AK103000.1 | - | 9 - 10 | 3302 | 75 | ||
More Information
Set Membership: |
|

LOC4333171
Genomic Map