Assay Details
| Gene Symbol: LOC4333171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775365.1 | XP_015630851.1 | 5 - 6 | 2281 | 93 | |
| XM_015775366.2 | XP_015630852.1 | 5 - 6 | 2259 | 93 | ||
| GenBank mRNA | AK065099.1 | - | 5 - 6 | 2110 | 93 | |
| AK067814.1 | - | 5 - 6 | 2231 | 93 | ||
| AK103000.1 | - | 2 - 3 | 1716 | 93 | ||
More Information
Set Membership: |
|

LOC4333171
Genomic Map