Assay Details
| Gene Symbol: LOC4333407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772504.2 | XP_015627990.1 | 1 - 2 | 233 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772504.2 transcripts.
Set Membership: |
|

LOC4333407
Genomic Map