Assay Details
Gene Symbol: LOC4333574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772441.2 | XP_015627927.1 | 6 - 7 | 792 | 130 | |
GenBank mRNA | AK064308.1 | - | 6 - 7 | 724 | 130 | |
AK103649.1 | - | 6 - 7 | 603 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
