Assay Details
| Gene Symbol: LOC4333678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772513.2 | XP_015627999.1 | 11 - 12 | 1329 | 71 | |
| XM_015772514.2 | XP_015628000.1 | 11 - 12 | 1329 | 71 | ||
| GenBank mRNA | AK067769.1 | - | 11 - 12 | 1332 | 71 | |
| AK099820.1 | - | 7 - 8 | 2825 | 71 | ||
More Information
Set Membership: |
|

LOC4333678
Genomic Map