Assay Details
Gene Symbol: LOC4333678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772513.2 | XP_015627999.1 | 12 - 13 | 1420 | 90 | |
GenBank mRNA | AK067769.1 | - | 12 - 13 | 1423 | 90 | |
AK099820.1 | - | 8 - 9 | 2916 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
