Assay Details
Gene Symbol: LOC4333678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772513.2 | XP_015627999.1 | 7 - 8 | 862 | 142 | |
XM_015772514.2 | XP_015628000.1 | 7 - 8 | 862 | 142 | ||
GenBank mRNA | AK067769.1 | - | 7 - 8 | 865 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
