Assay Details
| Gene Symbol: LOC4334276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775118.1 | XP_015630604.1 | 7 - 8 | 681 | 86 | |
| GenBank mRNA | AK065851.1 | - | 7 - 8 | 619 | 86 | |
| AK119532.1 | - | 7 - 8 | 671 | 86 | ||
| BT130523.1 | - | 4 - 5 | 322 | 86 | ||
More Information
Set Membership: |
|

LOC4334276
Genomic Map