Assay Details
| Gene Symbol: LOC4334312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775600.2 | XP_015631086.1 | 3 - 4 | 518 | 72 | |
| GenBank mRNA | AK100619.1 | - | 3 - 4 | 496 | 72 | |
More Information
Set Membership: |
|

LOC4334312
Genomic Map