Assay Details
| Gene Symbol: LOC4334314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776697.2 | XP_015632183.1 | 4 - 5 | 3024 | 71 | |
| XM_015776701.2 | XP_015632187.1 | 4 - 5 | 3267 | 71 | ||
| XM_015776702.2 | XP_015632188.1 | 5 - 6 | 3133 | 71 | ||
| GenBank mRNA | AK110946.1 | - | 3 - 4 | 386 | 71 | |
More Information
Set Membership: |
|

LOC4334314
Genomic Map