Assay Details
Gene Symbol: LOC4334752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774202.1 | XP_015629688.1 | 5 - 6 | 2605 | 99 | |
GenBank mRNA | AK070364.1 | - | 5 - 6 | 2577 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
