Assay Details
| Gene Symbol: LOC4335699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777903.1 | XP_015633389.1 | 6 - 7 | 744 | 73 | |
| GenBank mRNA | AK120062.1 | - | 6 - 7 | 750 | 73 | |
More Information
Set Membership: |
|

LOC4335699
Genomic Map