Assay Details
Gene Symbol: LOC4336045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780503.2 | XP_015635989.1 | 1 - 2 | 383 | 61 | |
GenBank mRNA | AK061516.1 | - | 1 - 2 | 289 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
