Assay Details
Gene Symbol: LOC4336045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780503.2 | XP_015635989.1 | 2 - 3 | 415 | 73 | |
GenBank mRNA | AK061516.1 | - | 2 - 3 | 321 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
