Assay Details
Gene Symbol: LOC4336045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780503.2 | XP_015635989.1 | 3 - 4 | 561 | 85 | |
XM_015780504.2 | XP_015635990.1 | 2 - 3 | 525 | 85 | ||
GenBank mRNA | AK061516.1 | - | 3 - 4 | 467 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
