Assay Details
Gene Symbol: LOC4336045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780503.2 | XP_015635989.1 | 4 - 5 | 638 | 59 | |
XM_015780504.2 | XP_015635990.1 | 3 - 4 | 602 | 59 | ||
GenBank mRNA | AK061516.1 | - | 4 - 5 | 544 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
