Assay Details
Gene Symbol: LOC4336045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780503.2 | XP_015635989.1 | 5 - 6 | 693 | 65 | |
XM_015780504.2 | XP_015635990.1 | 4 - 5 | 657 | 65 | ||
GenBank mRNA | AK061516.1 | - | 5 - 6 | 599 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
