Assay Details
| Gene Symbol: LOC4336195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779888.2 | XP_015635374.1 | 2 - 3 | 366 | 142 | |
| XM_015779889.2 | XP_015635375.1 | 2 - 3 | 366 | 142 | ||
| GenBank mRNA | AK111072.1 | - | 2 - 3 | 328 | 142 | |
More Information
Set Membership: |
|

LOC4336195
Genomic Map