Assay Details
Gene Symbol: LOC4336370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780188.2 | XP_015635674.1 | 10 - 11 | 2119 | 70 | |
XM_015780189.2 | XP_015635675.1 | 10 - 11 | 2046 | 70 | ||
GenBank mRNA | AK072655.1 | - | 10 - 11 | 2052 | 70 | |
AK103972.1 | - | 4 - 5 | 446 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
