Assay Details
Gene Symbol: LOC4336370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780188.2 | XP_015635674.1 | 11 - 12 | 2253 | 99 | |
XM_015780189.2 | XP_015635675.1 | 11 - 12 | 2180 | 99 | ||
GenBank mRNA | AK103972.1 | - | 5 - 6 | 580 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
