Assay Details
| Gene Symbol: LOC4336370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780188.2 | XP_015635674.1 | 13 - 14 | 2487 | 68 | |
| XM_015780189.2 | XP_015635675.1 | 13 - 14 | 2414 | 68 | ||
| GenBank mRNA | AK072655.1 | - | 13 - 14 | 2420 | 68 | |
| AK103972.1 | - | 7 - 8 | 814 | 68 | ||
More Information
Set Membership: |
|

LOC4336370
Genomic Map