Assay Details
Gene Symbol: LOC4336370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780188.2 | XP_015635674.1 | 1 - 2 | 290 | 87 | |
XM_015780189.2 | XP_015635675.1 | 1 - 2 | 262 | 87 | ||
GenBank mRNA | AK072655.1 | - | 1 - 2 | 268 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
