Assay Details
Gene Symbol: LOC4336370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780188.2 | XP_015635674.1 | 4 - 5 | 1030 | 87 | |
XM_015780189.2 | XP_015635675.1 | 4 - 5 | 957 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
