Assay Details
Gene Symbol: LOC4336370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780188.2 | XP_015635674.1 | 8 - 9 | 1887 | 80 | |
XM_015780189.2 | XP_015635675.1 | 8 - 9 | 1814 | 80 | ||
GenBank mRNA | AK072655.1 | - | 8 - 9 | 1820 | 80 | |
AK103972.1 | - | 2 - 3 | 214 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
