Assay Details
Gene Symbol: LOC4336370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780188.2 | XP_015635674.1 | 9 - 10 | 2013 | 79 | |
XM_015780189.2 | XP_015635675.1 | 9 - 10 | 1940 | 79 | ||
GenBank mRNA | AK072655.1 | - | 9 - 10 | 1946 | 79 | |
AK103972.1 | - | 3 - 4 | 340 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
