Assay Details
| Gene Symbol: LOC4336722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781552.1 | XP_015637038.1 | 11 - 12 | 3183 | 67 | |
| XM_015781553.2 | XP_015637039.1 | 13 - 14 | 3459 | 67 | ||
| XM_015781554.2 | XP_015637040.1 | 13 - 14 | 3456 | 67 | ||
| XM_015781555.2 | XP_015637041.1 | 13 - 14 | 3456 | 67 | ||
| XM_015781556.2 | XP_015637042.1 | 13 - 14 | 3453 | 67 | ||
| XM_015781557.2 | XP_015637043.1 | 9 - 10 | 2807 | 67 | ||
| GenBank mRNA | AK064589.1 | - | 1 - 2 | 120 | 67 | |
More Information
Set Membership: |
|

LOC4336722
Genomic Map