Assay Details
| Gene Symbol: LOC4336722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781552.1 | XP_015637038.1 | 12 - 13 | 3326 | 114 | |
| XM_015781553.2 | XP_015637039.1 | 14 - 15 | 3602 | 114 | ||
| XM_015781554.2 | XP_015637040.1 | 14 - 15 | 3599 | 114 | ||
| XM_015781555.2 | XP_015637041.1 | 14 - 15 | 3599 | 114 | ||
| XM_015781556.2 | XP_015637042.1 | 14 - 15 | 3596 | 114 | ||
| XM_015781557.2 | XP_015637043.1 | 10 - 11 | 2950 | 114 | ||
| GenBank mRNA | AK064589.1 | - | 2 - 3 | 263 | 114 | |
More Information
Set Membership: |
|

LOC4336722
Genomic Map