Assay Details
| Gene Symbol: LOC4336722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781552.1 | XP_015637038.1 | 15 - 16 | 3865 | 114 | |
| XM_015781553.2 | XP_015637039.1 | 17 - 18 | 4141 | 114 | ||
| XM_015781554.2 | XP_015637040.1 | 17 - 18 | 4138 | 114 | ||
| XM_015781555.2 | XP_015637041.1 | 17 - 18 | 4138 | 114 | ||
| XM_015781556.2 | XP_015637042.1 | 17 - 18 | 4135 | 114 | ||
| XM_015781557.2 | XP_015637043.1 | 13 - 14 | 3489 | 114 | ||
| GenBank mRNA | AK064589.1 | - | 5 - 6 | 802 | 114 | |
More Information
Set Membership: |
|

LOC4336722
Genomic Map