Assay Details
| Gene Symbol: LOC4336755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780890.1 | XP_015636376.1 | 6 - 7 | 1698 | 75 | |
| GenBank mRNA | AK072398.1 | - | 6 - 7 | 1704 | 75 | |
| AK104058.1 | - | 1 - 2 | 64 | 75 | ||
More Information
Set Membership: |
|

LOC4336755
Genomic Map