Assay Details
| Gene Symbol: LOC4337088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780662.2 | XP_015636148.1 | 14 - 15 | 1444 | 127 | |
| GenBank mRNA | AK065665.1 | - | 14 - 15 | 1451 | 127 | |
| AK099320.1 | - | 14 - 15 | 1401 | 127 | ||
More Information
Set Membership: |
|

LOC4337088
Genomic Map