Assay Details
| Gene Symbol: LOC4337088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780662.2 | XP_015636148.1 | 3 - 4 | 407 | 120 | |
| GenBank mRNA | AK065665.1 | - | 3 - 4 | 414 | 120 | |
| AK099320.1 | - | 3 - 4 | 364 | 120 | ||
More Information
Set Membership: |
|

LOC4337088
Genomic Map