Assay Details
Gene Symbol: LOC4337338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777975.2 | XP_015633461.1 | 1 - 2 | 304 | 153 | |
GenBank mRNA | AK288046.1 | - | 1 - 2 | 164 | 153 | |
AY224471.1 | - | 1 - 2 | 128 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
