Assay Details
Gene Symbol: LOC4337338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777975.2 | XP_015633461.1 | 2 - 3 | 435 | 134 | |
GenBank mRNA | AK288046.1 | - | 2 - 3 | 295 | 134 | |
AY224471.1 | - | 2 - 3 | 259 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
