Assay Details
Gene Symbol: LOC4337338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777975.2 | XP_015633461.1 | 4 - 5 | 599 | 125 | |
GenBank mRNA | AK288046.1 | - | 4 - 5 | 459 | 125 | |
AY224471.1 | - | 4 - 5 | 423 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
