Assay Details
Gene Symbol: LOC4337338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777975.2 | XP_015633461.1 | 7 - 8 | 770 | 120 | |
GenBank mRNA | AK288046.1 | - | 7 - 8 | 630 | 120 | |
AY224471.1 | - | 7 - 8 | 594 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
