Assay Details
| Gene Symbol: LOC4337720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782361.2 | XP_015637847.1 | 1 - 2 | 297 | 117 | |
| GenBank mRNA | AB119279.1 | - | 1 - 2 | 239 | 117 | |
| AK068407.1 | - | 1 - 2 | 274 | 117 | ||
| AK099152.1 | - | 1 - 2 | 274 | 117 | ||
More Information
Set Membership: |
|

LOC4337720
Genomic Map