Assay Details
Gene Symbol: LOC4337994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782186.2 | XP_015637672.1 | 11 - 12 | 1524 | 143 | |
GenBank mRNA | AK102592.1 | - | 11 - 12 | 1536 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
