Assay Details
| Gene Symbol: LOC4338246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782515.2 | XP_015638001.1 | 3 - 4 | 571 | 81 | |
| XM_015782516.2 | XP_015638002.1 | 4 - 5 | 867 | 81 | ||
| XM_015782518.2 | XP_015638004.1 | 3 - 4 | 559 | 81 | ||
| GenBank mRNA | AK065555.1 | - | 4 - 5 | 1060 | 81 | |
More Information
Set Membership: |
|

LOC4338246
Genomic Map