Assay Details
| Gene Symbol: LOC4338646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785117.1 | XP_015640603.1 | 2 - 3 | 497 | 95 | |
| GenBank mRNA | AK061728.1 | - | 2 - 3 | 502 | 95 | |
| AK065038.1 | - | 2 - 3 | 306 | 95 | ||
More Information
Set Membership: |
|

LOC4338646
Genomic Map