Assay Details
Gene Symbol: LOC4338988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782496.2 | XP_015637982.1 | 2 - 3 | 526 | 65 | |
XM_015782497.2 | XP_015637983.1 | 2 - 3 | 537 | 65 | ||
GenBank mRNA | AK103232.1 | - | 2 - 3 | 470 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
